9-Benzylcarbazole 99+%

Supplier: Ambeed
Warning

A499124-10G A499124-1G A499124-250MG A499124-25G A499124-5G
76789-902EA 250 USD
76789-902 76789-904 76789-906 76789-908 77669-242
9-Benzylcarbazole 99+%
9-Benzylcarbazole

Precaución: for Research Use Only

Formula: C₁₉H₁₅N
MDL Number: MFCD00218286
CAS Number: 19402-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR