Boc-(R)-3-amino-4-(3,4-difluorophenyl)butyric acid 98%

Supplier: Ambeed
Warning

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-4-(3,4-difluorophenyl)butanoic acid

A497717-100MG A497717-1G A497717-250MG
76789-890EA 45.62 USD
76789-890 76789-892 76789-894
Boc-(R)-3-amino-4-(3,4-difluorophenyl)butyric acid 98%
Boc-(R)-3-amino-4-(3,4-difluorophenyl)butyric acid

Precaución: for Research Use Only

Formula: C₁₅H₁₉F₂NO₄
MW: 315.32 g/mol
MDL Number: MFCD01860952
CAS Number: 269396-59-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR