Thiotepa 97%

Supplier: Ambeed
Danger

Synonyms: Tri(aziridin-1-yl)phosphine sulphide, Thio-TEPA, Tri(1-aziridinyl)phosphine sulphide

A496771-1G A496771-1MG A496771-250MG A496771-100MG
76789-880EA 377.69 USD
76789-880 76789-882 76789-884 77602-142
Thiotepa 97%
Thiotepa

Precaución: for Research Use Only

Formula: C₆H₁₂N₃PS
MW: 189.217 g/mol
MDL Number: MFCD00145452
CAS Number: 52-24-4
UN: 2811
ADR: 6.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR