2-Benzylbenzoic acid 98%

Supplier: Ambeed
Warning

A496385-100G A496385-10G A496385-5G A496385-25G A496385-500G
76789-874EA 131 USD
76789-874 76789-876 76789-878 77602-138 77669-228
2-Benzylbenzoic acid 98%
2-Benzylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₂
MDL Number: MFCD00002484
CAS Number: 612-35-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR