5-Bromo-4-methyl-1H-indazole 98%

Supplier: Ambeed
Warning

A495393-100MG A495393-250MG A495393-5G A495393-10G
76789-860EA 6.37 USD
76789-860 76789-862 76789-864 77602-136
5-Bromo-4-methyl-1H-indazole 98%
5-Bromo-4-methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
MDL Number: MFCD11007842
CAS Number: 1082041-34-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR