N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl (tert-butoxycarbonyl)-L-phenylalaninate, N-Boc-L-phenylalanine Methyl Ester, Boc-Phe-OMe
A495178-100G
A495178-10G
A495178-25G
A495178-5G
76789-852EA
65
USD
76789-852
76789-854
76789-856
76789-858
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester 97%
N-(tert-Butoxycarbonyl)-L-phenylalanine methyl ester
Precaución: for Research Use Only
Formula:
C₁₅H₂₁NO₄ MW: 279.34 g/mol |
MDL Number:
MFCD00076977 CAS Number: 51987-73-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...