3-Methyldihydro-2H-pyran-4(3H)-one 98%

Supplier: Ambeed
Warning

A493849-100MG A493849-1G A493849-250MG A493849-5G
76789-836EA 36 USD
76789-836 76789-838 76789-840 76789-842
3-Methyldihydro-2H-pyran-4(3H)-one 98%
3-Methyldihydro-2H-pyran-4(3H)-one

Precaución: for Research Use Only

Formula: C6H10O2
MDL Number: MFCD13193744
CAS Number: 119124-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR