Dehydrocostus Lactone 98%

Supplier: Ambeed
Warning

A493627-1MG A493627-5MG A493627-10MG A493627-5G A493627-100MG A493627-250MG A493627-1G
76789-832EA 7 USD
76789-832 76789-834 77602-130 77669-214 77669-210 77669-212 77872-819
Dehydrocostus Lactone 98%
Dehydrocostus Lactone

Precaución: for Research Use Only

Formula: C₁₅H₁₈O₂
MDL Number: MFCD00210277
CAS Number: 477-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR