5-Bromo-2-fluoro-4-methylbenzaldehyde 98%

Supplier: Ambeed
Warning

A493302-1G A493302-5G
76789-822EA 32 USD
76789-822 77872-818
5-Bromo-2-fluoro-4-methylbenzaldehyde 98%
5-Bromo-2-fluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD04112582
CAS Number: 497224-12-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR