4-Methoxycyclohexanecarboxylic acid 98%

Supplier: Ambeed
Warning

A491690-25G A491690-5G A491690-1G
76789-808EA 815.84 USD
76789-808 76789-810 77669-204
4-Methoxycyclohexanecarboxylic acid 98%
4-Methoxycyclohexanecarboxylic acid

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.2 g/mol
MDL Number: MFCD00074992
CAS Number: 95233-12-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR