3-Iodo-5-(trifluoromethyl)-2-pyridinylamine 97%

Supplier: Ambeed
Warning

Synonyms: 3-Iodo-5-(trifluoromethyl)pyridin-2-amine, 2-Amino-3-iodo-5-(trifluoromethyl)pyridine

A491517-100MG A491517-10G A491517-5G A491517-1G
76789-802EA 18.04 USD
76789-802 76789-804 76789-806 77669-202
3-Iodo-5-(trifluoromethyl)-2-pyridinylamine 97%
3-Iodo-5-(trifluoromethyl)-2-pyridinylamine

Precaución: for Research Use Only

Formula: C₆H₄F₃IN₂
MW: 288.01 g/mol
MDL Number: MFCD10697686
CAS Number: 211308-82-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR