4-(4-Ethylpiperazin-1-yl)phenylboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

A491162-100MG A491162-1G A491162-250MG A491162-5G
76789-796EA 6 USD
76789-796 76789-798 76789-800 77872-816
4-(4-Ethylpiperazin-1-yl)phenylboronic acid pinacol ester 95%
4-(4-Ethylpiperazin-1-yl)phenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₉BN₂O₂
MDL Number: MFCD13190738
CAS Number: 656257-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR