4-Bromo-3-fluorophenylacetic acid 98%

Supplier: Ambeed
Warning

A490096-100G A490096-10G A490096-1G A490096-250MG A490096-25G A490096-500G A490096-5G
76789-782EA 411.63 USD
76789-782 76789-784 76789-786 76789-788 76789-790 76789-792 76789-794
4-Bromo-3-fluorophenylacetic acid 98%
4-Bromo-3-fluorophenylacetic acid

Precaución: for Research Use Only

Formula: C₈H₆BrFO₂
MW: 233.0300 g/mol
MDL Number: MFCD12756200
CAS Number: 942282-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR