3-(Boc-amino)-3-phenylpropionic acid 98%

Supplier: Ambeed
Warning

Synonyms: Boc-DL-β-Phe-OH, 3-tert-Butoxycarbonylamino-3-phenyl-propionic acid

A490094-1G A490094-25G A490094-5G
76789-776EA 15.92 USD
76789-776 76789-778 76789-780
3-(Boc-amino)-3-phenylpropionic acid 98%
3-(Boc-amino)-3-phenylpropionic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₉NO₄
MW: 265.31 g/mol
MDL Number: MFCD02090695
CAS Number: 14676-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR