Z-Gln-Gly-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Benzyloxycarbonyl-L-glutaminylglycine, (S)-2-(5-Amino-2-(((benzyloxy)carbonyl)amino)-5-oxopentanamido)acetic acid

A488647-1G A488647-25G A488647-5G A488647-250MG
76789-768EA 13 USD
76789-768 76789-770 76789-772 77669-190
Z-Gln-Gly-OH 98%
Z-Gln-Gly-OH

Precaución: for Research Use Only

Formula: C₁₅H₁₉N₃O₆
MW: 337.332 g/mol
MDL Number: MFCD00055926
CAS Number: 6610-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR