6-Amino-3-bromo-2,4-dimethylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-4,6-dimethylpyridin-2-amine, 3-Bromo-2,4-dimethylpyridin-6-amine

A488112-1G A488112-250MG A488112-5G
76789-758EA 33.95 USD
76789-758 76789-760 76789-762
6-Amino-3-bromo-2,4-dimethylpyridine 98%
6-Amino-3-bromo-2,4-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MW: 201.064 g/mol
MDL Number: MFCD00151793
CAS Number: 89856-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR