Z-Lys(Boc)-ONp 95%

Supplier: Ambeed
Warning

A486512-10G A486512-1G A486512-25G A486512-5G
76789-732EA 25 USD
76789-732 76789-734 76789-736 76789-738
Z-Lys(Boc)-ONp 95%
Z-Lys(Boc)-ONp

Precaución: for Research Use Only

Formula: C₂₅H₃₁N₃O₈
MDL Number: MFCD00076963
CAS Number: 2212-69-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR