Ethyl-6-(trifluoromethyl)nicotinate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 6-(trifluoromethyl)nicotinate

A486440-100G A486440-1G A486440-25G A486440-5G
76789-724EA 482.71 USD
76789-724 76789-726 76789-728 76789-730
Ethyl-6-(trifluoromethyl)nicotinate 98%
Ethyl-6-(trifluoromethyl)nicotinate

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD04972865
CAS Number: 597532-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR