2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate, 2-Acetoxyphenylboronic acid pinacol ester
A485136-1G
A485136-250MG
A485136-5G
76789-710EA
98
USD
76789-710
76789-712
76789-714
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate 98%
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl acetate
Precaución: for Research Use Only
Formula:
C₁₄H₁₉BO₄ |
MDL Number:
MFCD03453062 CAS Number: 480424-68-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...