2-(Benzyloxy)benzoic acid 98%

Supplier: Ambeed
Warning

A480620-1G A480620-25G A480620-5G A480620-10G
76789-674EA 9 USD
76789-674 76789-676 76789-678 77872-793
2-(Benzyloxy)benzoic acid 98%
2-(Benzyloxy)benzoic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₂O₃
MDL Number: MFCD00051940
CAS Number: 14389-86-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR