4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Nitrobenzo-15-crown-5
A480372-1G
A480372-25G
A480372-5G
76789-672EA
56
USD
76789-672
77872-791
77872-792
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane) 99%
4'-Nitrobenzo-15-crown-5 (4'-nitrobenzo-1,4,7,10,13-pentaoxacyclopentadecane)
Precaución: for Research Use Only
Formula:
C₁₄H₁₉NO₇ MW: 313.30 g/mol |
MDL Number:
MFCD00060713 CAS Number: 60835-69-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...