2'-(Trifluoromethyl)biphenyl-3-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
2'-(Trifluoromethyl)-[1,1'-biphenyl]-3-carboxylic acid
A480352-1G
A480352-250MG
A480352-5G
76789-666EA
107.16
USD
76789-666
76789-668
76789-670
2'-(Trifluoromethyl)biphenyl-3-carboxylic acid 97%
2'-(Trifluoromethyl)biphenyl-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₄H₉F₃O₂ MW: 266.22 g/mol |
MDL Number:
MFCD03424602 CAS Number: 168618-48-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...