[(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride 98%

Supplier: Ambeed
Warning

Synonyms: Dichloro[(R)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II), Dichloro[(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II), Dichloro [(R)-(+)-2,2'-bis(diphenylphosphino)-1,1'-binaphathyl] ruthenium(II)

A480159-100MG A480159-1G A480159-250MG A480159-5G
76789-658EA 8 USD
76789-658 76789-660 76789-662 76789-664
[(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride 98%
[(R)-2,2'-Bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)dichloride

Precaución: for Research Use Only

Formula: C₄₄H₃₂Cl₂P₂Ru
MW: 794.66 g/mol
MDL Number: MFCD01073794
CAS Number: 132071-87-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR