4-Methoxypyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A479850-100MG A479850-1G
76789-654EA 18 USD
76789-654 76789-656
4-Methoxypyridin-2(1H)-one 95%
4-Methoxypyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MDL Number: MFCD09839191
CAS Number: 52545-13-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR