2-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)thiazole-4-carboxamide 98%

Supplier: Ambeed
Warning

A479093-10MG A479093-1MG A479093-5MG
76789-648EA 83.82 USD
76789-648 76789-650 76789-652
2-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)thiazole-4-carboxamide 98%
2-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)thiazole-4-carboxamide

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₅S
MW: 260.26 g/mol
MDL Number: MFCD00866494
CAS Number: 60084-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR