Benzene-1,3,5-tricarbonyl trichloride 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
1,3,5-Benzenetricarboxylic acid chloride, 1,3,5-Benzenetricarbonyl chloride, Trimesoylchloride, Trimesoyl chloride
A478592-25G
A478592-5G
A478592-100G
76789-642EA
10.61
USD
76789-642
76789-644
76789-646
77669-148
Benzene-1,3,5-tricarbonyl trichloride 98%
Benzene-1,3,5-tricarbonyl trichloride
Precaución: for Research Use Only
Formula:
C₉H₃Cl₃O₃ MW: 265.48 g/mol Boiling Pt: 312 |
MDL Number:
MFCD00000679 CAS Number: 4422-95-1 UN: 3261 ADR: 8 Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...