3,3-Diphenylpropylamine 98%

Supplier: Ambeed
Warning

A476787-100G A476787-1G A476787-25G A476787-5G
76789-628EA 98 USD
76789-628 76789-630 76789-632 76789-634
3,3-Diphenylpropylamine 98%
3,3-Diphenylpropylamine

Precaución: for Research Use Only

Formula: C₁₅H₁₇N
MDL Number: MFCD00008202
CAS Number: 5586-73-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR