Tetramethylammonium hydrogen sulfate monohydrate 98%

Supplier: Ambeed
Warning

A476261-100G A476261-25G A476261-5G
76789-622EA 76 USD
76789-622 76789-624 76789-626
Tetramethylammonium hydrogen sulfate monohydrate 98%
Tetramethylammonium hydrogen sulphate monohydrate

Precaución: for Research Use Only

Formula: C₄H₁₃NO₄S·1H₂O
MDL Number: MFCD00150356
CAS Number: 103812-00-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR