2-Hydroxy-5-iodo-3-(trifluoromethyl)pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Iodo-3-(trifluoromethyl)pyridin-2-ol

A475877-1G A475877-250MG A475877-5G
76789-616EA 11 USD
76789-616 76789-618 76789-620
2-Hydroxy-5-iodo-3-(trifluoromethyl)pyridine 95%
2-Hydroxy-5-iodo-3-(trifluoromethyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₃F₃INO
MW: 288.994 g/mol
MDL Number: MFCD12024303
CAS Number: 887707-23-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR