1-(4-Bromophenyl)ethanamine hydrochloride 98%

Supplier: Ambeed
Warning

A475402-1G A475402-25G A475402-5G
76789-610EA 22.28 USD
76789-610 76789-612 76789-614
1-(4-Bromophenyl)ethanamine hydrochloride 98%
1-(4-Bromophenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₁BrClN
MW: 236.53 g/mol
MDL Number: MFCD06662255
CAS Number: 90006-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR