1-(4-Isopropoxyphenyl)methanamine 98%

Supplier: Ambeed
Danger

Synonyms: 4-Isopropoxybenzylamine

A475336-1G A475336-250MG A475336-5G
76789-604EA 54 USD
76789-604 76789-606 76789-608
1-(4-Isopropoxyphenyl)methanamine 98%
1-(4-Isopropoxyphenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₅NO
MW: 165.24 g/mol
MDL Number: MFCD04037892
CAS Number: 21244-34-8
UN: 2735
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR