Methyl diphenylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl Diphenylacetate

A473909-25G A473909-5G A473909-100G
76789-596EA 39.26 USD
76789-596 76789-598 77669-134
Methyl diphenylacetate 98%
Methyl diphenylacetate

Precaución: for Research Use Only

Formula: C₁₅H₁₄O₂
MW: 226.28 g/mol
MDL Number: MFCD00025869
CAS Number: 3469-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR