3-Iodo-o-toluic acid 98%

Supplier: Ambeed
Warning

A472495-100G A472495-1G A472495-25G A472495-5G
76789-578EA 116 USD
76789-578 76789-580 76789-582 76789-584
3-Iodo-o-toluic acid 98%
3-Iodo-o-toluic acid

Precaución: for Research Use Only

Formula: C₈H₇IO₂
MDL Number: MFCD00270112
CAS Number: 133232-56-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR