Thiophene-2-carboxaldoxime 98%

Supplier: Ambeed
Danger

A469888-25G A469888-5G
76789-558EA 105.03 USD
76789-558 76789-560
Thiophene-2-carboxaldoxime 98%
Thiophene-2-carboxaldoxime

Precaución: for Research Use Only

Formula: C₅H₅NOS
MW: 127.17 g/mol
MDL Number: MFCD00022509
CAS Number: 29683-84-9
UN: 1325
ADR: 4.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR