Benzyl-N-[1-carbamoyl-2-(4-hydroxyphenyl)ethyl]carbamate 98%

Supplier: Ambeed
Warning

Synonyms: Z-Tyr-NH₂, (S)-Benzyl (1-amino-3-(4-hydroxyphenyl)-1-oxopropan-2-yl)carbamate

A469807-1G A469807-250MG
76789-554EA 40 USD
76789-554 76789-556
Benzyl-N-[1-carbamoyl-2-(4-hydroxyphenyl)ethyl]carbamate 98%
Benzyl-N-[1-carbamoyl-2-(4-hydroxyphenyl)ethyl]carbamate

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₂O₄
MW: 314.341 g/mol
MDL Number: MFCD00136595
CAS Number: 19898-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR