3-Bromo-2-methylthiophene 95%
Supplier: Ambeed
|
Danger
|
A468362-5G
A468362-25G
A468362-1G
76789-550EA
88
USD
76789-550
77669-106
77872-779
3-Bromo-2-methylthiophene 95%
3-Bromo-2-methylthiophene
Precaución: for Research Use Only
Formula:
C₅H₅BrS |
MDL Number:
MFCD09033008 CAS Number: 30319-05-2 UN: 1992 ADR: 3,6.1,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...