4-Chloropyrrolo[1,2-f][1,2,4]triazine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Chloropyrrolo[2,1-f][1,2,4]triazine

A466850-100MG A466850-5G A466850-1G
76789-546EA 44 USD
76789-546 76789-548 77669-102
4-Chloropyrrolo[1,2-f][1,2,4]triazine 95%
4-Chloropyrrolo[1,2-f][1,2,4]triazine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MW: 153.57 g/mol
MDL Number: MFCD11519387
CAS Number: 888720-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR