(2S,3R,4S,5S,6R)-2-(((6aR,12aR)-6a,12a-Dihydro-6H-[1,3]dioxolo[4',5':5,6]benzofuro[3,2-c]chromen-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 97%

Supplier: Ambeed
Warning

Synonyms: Trifolirhizin

A466656-10MG A466656-1MG A466656-5MG
76789-540EA 68 USD
76789-540 76789-542 76789-544
(2S,3R,4S,5S,6R)-2-(((6aR,12aR)-6a,12a-Dihydro-6H-[1,3]dioxolo[4',5':5,6]benzofuro[3,2-c]chromen-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol 97%
(2S,3R,4S,5S,6R)-2-(((6aR,12aR)-6a,12a-Dihydro-6H-[1,3]dioxolo[4',5':5,6]benzofuro[3,2-c]chromen-3-yl)oxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

Precaución: for Research Use Only

Formula: C₂₂H₂₂O₁₀
MW: 446.4 g/mol
MDL Number: MFCD00238700
CAS Number: 6807-83-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR