Tripropyl orthoformate 95%

Supplier: Ambeed
Warning

Synonyms: 1,1',1''-[Methylidynetris(oxy)]trispropane, Tri-n-propyl orthoformate, Tripropylorthoformate

A466010-100G A466010-1G A466010-25G A466010-5G
76789-532EA 262 USD
76789-532 76789-534 76789-536 76789-538
Tripropyl orthoformate 95%
Tripropyl orthoformate

Precaución: for Research Use Only

Formula: C₁₀H₂₂O₃
MDL Number: MFCD00015214
CAS Number: 621-76-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR