(R)-Benzyl (2-oxotetrahydrofuran-3-yl)carbamate 98+%

Supplier: Ambeed
Warning

A465929-1G A465929-250MG A465929-5G
76789-524EA 25 USD
76789-524 76789-526 76789-528
(R)-Benzyl (2-oxotetrahydrofuran-3-yl)carbamate 98+%
(R)-Benzyl (2-oxotetrahydrofuran-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₄
MW: 235.23 g/mol
MDL Number: MFCD11519127
CAS Number: 41088-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR