Tripropyl phosphate 98%

Supplier: Ambeed
Warning

A465857-100G A465857-5G A465857-25G
76789-520EA 174 USD
76789-520 76789-522 77872-776
Tripropyl phosphate 98%
Tripropyl phosphate

Precaución: for Research Use Only

Formula: C₉H₂₁O₄P
MW: 224.2344 g/mol
MDL Number: MFCD00009368
CAS Number: 513-08-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR