Zinc trifluoromethanesulphonate 98%

Supplier: Ambeed
Danger

Synonyms: Zinc(II) trifluoromethanesulphonate, Zinc triflate, Zinc trifluoromethanesulphonate

A465167-10G A465167-25G A465167-5G
76789-506EA 24 USD
76789-506 76789-508 76789-510
Zinc trifluoromethanesulphonate 98%
Zinc trifluoromethanesulphonate

Precaución: for Research Use Only

Formula: C₂F₆O₆S₂Zn
MW: 363.53
MDL Number: MFCD00013229
CAS Number: 54010-75-2
UN: 3261
ADR: 8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR