5-Bromo-4-chloro-1H-indazole 98%

Supplier: Ambeed
Danger

A463746-100MG A463746-1G
76789-496EA 10.61 USD
76789-496 76789-498
5-Bromo-4-chloro-1H-indazole 98%
5-Bromo-4-chloro-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₄BrClN₂
MW: 231.4771 g/mol
MDL Number: MFCD11007933
CAS Number: 1082041-90-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR