(1-(tert-Butoxycarbonyl)-6-fluoro-1H-indol-2-yl)boronic acid 98%

Supplier: Ambeed
Warning

A460882-1G A460882-250MG A460882-100MG
76789-474EA 193.09 USD
76789-474 77601-996 77669-070
(1-(tert-Butoxycarbonyl)-6-fluoro-1H-indol-2-yl)boronic acid 98%
(1-(tert-Butoxycarbonyl)-6-fluoro-1H-indol-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₅BFNO₄
MW: 279.0719 g/mol
MDL Number: MFCD09953519
CAS Number: 1000068-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR