trans-Di(mu-acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II) 98%

Supplier: Ambeed
Warning

Synonyms: trans-Di-μ-acetatobis[2-[bis(2-methylphenyl)phosphino]benzyl]dipalladium, cataCXium® C

A460830-1G A460830-100MG A460830-250MG A460830-5G
76789-472EA 46 USD
76789-472 77669-068 77872-764 77872-765
trans-Di(mu-acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II) 98%
trans-Di(mu-acetato)bis[o-(di-o-tolylphosphino)benzyl]dipalladium(II)

Precaución: for Research Use Only

Formula: C₄₆H₄₆O₄P₂Pd₂
MW: 937.6400 g/mol
MDL Number: MFCD01075746
CAS Number: 172418-32-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR