tert-Pentylbenzene 97%

Supplier: Ambeed
Danger

A460774-100G A460774-500G
76789-468EA 24 USD
76789-468 76789-470
tert-Pentylbenzene 97%
tert-Pentylbenzene

Precaución: for Research Use Only

Formula: C₆H₅C(CH₃)₂CH₂CH₃
MDL Number: MFCD00015187
CAS Number: 2049-95-8
UN: 3295
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR