4-Iodo-1H-indazole 95%

Supplier: Ambeed
Warning

A459698-250MG A459698-1G
76789-456EA 50.93 USD
76789-456 77669-064
4-Iodo-1H-indazole 95%
4-Iodo-1H-indazole

Precaución: for Research Use Only

Formula: C₇H₅IN₂
MW: 244.0300 g/mol
MDL Number: MFCD07781607
CAS Number: 885522-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR