Tris(4-methoxy-3,5-dimethylphenyl)phosphine 98%
Supplier: Ambeed
|
Warning
|
A458096-250MG
A458096-5G
76789-452EA
41
USD
76789-452
77669-062
Tris(4-methoxy-3,5-dimethylphenyl)phosphine 98%
Tris(4-methoxy-3,5-dimethylphenyl)phosphine
Precaución: for Research Use Only
Formula:
C₂₇H₃₃O₃P |
MDL Number:
MFCD03840342 CAS Number: 121898-64-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...