Tizanidine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)benzo[c][1,2,5]thiadiazol-4-amine hydrochloride, Tizanidine HCl, 5-Chloro-4-[(4,5-dihydro-1H-imidazol-2-yl)amino]-2,1,3-benzothiadiazole Hydrochloride

A458091-100MG A458091-1G A458091-25G A458091-5G A458091-100G A458091-50MG
76789-444EA 5.31 USD
76789-444 76789-446 76789-448 76789-450 77669-058 77669-060
Tizanidine hydrochloride 98%
Tizanidine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₈ClN₅S
MW: 290.17 g/mol
MDL Number: MFCD00798231
CAS Number: 64461-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR