Strontium carbonate 99%

Supplier: Ambeed
Warning

A456957-1000G A456957-100G A456957-25G A456957-500G
76789-428EA 17 USD
76789-428 76789-430 76789-432 76789-434
Strontium carbonate 99%
Strontium carbonate

Precaución: for Research Use Only

Formula: CO₃Sr
MDL Number: MFCD00011250
CAS Number: 1633-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR